Skip to Content
MilliporeSigma

Skip To

XPIH9BD00E9A

AiFChem (an XtalPi company)

N,N,N,N-Tetramethyl-1,4-phenylenediamine

Synonym(s):

N~1~,N~1~,N~4~,N~4~-tetramethyl-1,4-benzenediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C10H16N2
CAS Number:
Molecular Weight:
164.25
MDL number:
Product XPIH9BD00E9A is currently not available for purchase. Contact Technical Service


Inchi Code

1S/C10H16N2/c1-11(2)9-5-7-10(8-6-9)12(3)4/h5-8H,1-4H3

Inchi Key

CJAOGUFAAWZWNI-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Physical Form

Form: solid or semi-solid or liquid

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Keep in dark place,Inert atmosphere,2-8C

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Keep in dark place,Inert atmosphere,2-8C



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library