Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

XPIH9BD095D8

AiFChem (an XtalPi company)

Ethyl 3-hydrazinylbenzoate hydrochloride

Synonym(s):

ethyl 3-hydrazinobenzoate hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product XPIH9BD095D8 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C9H13O2N2Cl1
CAS Number:
Molecular Weight:
216.67
MDL number:

Skip To

Inchi Code

1S/C9H12N2O2.ClH/c1-2-13-9(12)7-4-3-5-8(6-7)11-10;/h3-6,11H,2,10H2,1H3;1H

Inchi Key

YMKGDNLSBACMSE-UHFFFAOYSA-N

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Keep in dark place,Inert atmosphere,Room temperature

Purity

95%

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Keep in dark place,Inert atmosphere,Room temperature

Country of Origin

CN

Physical Form

Form: solid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library