Skip to Content
MilliporeSigma

XPIH9BD09AAD

AiFChem (an XtalPi company)

6-Quinolinecarboxaldehyde

Synonym(s):

6-quinolinecarbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H7O1N1
CAS Number:
Molecular Weight:
157.17
MDL number:


Inchi Code

1S/C10H7NO/c12-7-8-3-4-10-9(6-8)2-1-5-11-10/h1-7H

Inchi Key

VUAOIXANWIFYCU-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Physical Form

White to pale-yellow to yellow to orange powder or crytals

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Inert atmosphere,2-8C

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully@resealed and kept upright to prevent leakage.Inert atmosphere,2-8C



Still not finding the right product?

Explore all of our products under 6-Quinolinecarboxaldehyde


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library