Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

XPIH9BD0E614

AiFChem (an XtalPi company)

2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

Synonym(s):

2-{(E)-[4-(dimethylamino)phenyl]diazenyl}benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

500 G

$62.00

1 KG

$123.00

Vendor SKU: ACWMCV106-500G

$62.00


Ships in 7 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C15H15N3O2/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-10H,1-2H3,(H,19,20)

Inchi Key

CEQFOVLGLXCDCX-UHFFFAOYSA-N

Shipping Temperature

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Keep in dark place,Sealed in dry,Room Temperature

Purity

4.56.2

Storage temp.

Keep container tightly closed in a dry and well-ventilated area. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Keep in dark place,Sealed in dry,Room Temperature

Country of Origin

CN

Physical Form

Red to Deep red to Red brown Powder or crystals


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service