bp
104 C
density
0.883
Iupac Name
pentyl nitrite
Inchi Code
1S/C5H11NO2/c1-2-3-4-5-8-6-7/h2-5H2,1H3
InChI key
CSDTZUBPSYWZDX-UHFFFAOYSA-N
storage temp.
Store at 2-8C,Argon charged
Purity
95%
Country of Origin
US
Shipping Temperature
Wet ice
Product Link
https://www.aladdinsci.com/amyl-nitrite-bioactive-small-molecules-by-aladdin-scientific-a151126.html
Flash Point
-18C(lit.)
Boiling point
104 C
density
0.883
Signal Word
Danger
Hazard Statements
MSDS
https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/A151126-SCI_c0523124280912dcf28e92b1e3857536.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service