Skip to Content
MilliporeSigma

ALNH9A9D6B74

Aladdin Scientific

3-(Aminomethyl)piperidine

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
114.19
MDL number:

bp

89C/18mmHg(lit.)

Iupac Name

3-piperidinylmethanamine

Inchi Code

1S/C6H14N2/c7-4-6-2-1-3-8-5-6/h6,8H,1-5,7H2

InChI key

IPOVLZSJBYKHHU-UHFFFAOYSA-N

storage temp.

Store at -20C,Argon charged

Purity

98%

Country of Origin

US

Shipping Temperature

Ice chest + Ice pads

Physical Form

Liquid

Product Link

https://www.aladdinsci.com/3-aminomethyl-piperidine-bioactive-small-molecules-by-aladdin-scientific-a151610.html

Flash Point

77C(lit.)

Boiling point

89C/18mmHg(lit.)

pictograms

Corrosion

Signal Word

Danger

Hazard Statements

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/A151610-SCI_10a67174442feb0d3f9aa1aafc1708fc.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library