Skip to Content
MilliporeSigma

ALNH9A9DB378

Aladdin Scientific

3-Bromophenol

Synonym(s):

3-bromophenol

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
173.01
MDL number:

bp

236C

mp

30-32C

Inchi Code

1S/C6H5BrO/c7-5-2-1-3-6(8)4-5/h1-4,8H

Inchi Key

MNOJRWOWILAHAV-UHFFFAOYSA-N

Storage temp.

Argon charged

Purity

98%

Country of Origin

US

Shipping Temperature

Normal

Flash Point

>110C

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/B106994-SCI_486c40c61bee4ce736df8a681a5a1ad9.pdf

signalword

Warning

pictograms

EnvironmentExclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library