Skip to Content
MilliporeSigma

ALNH9A9DB53F

Aladdin Scientific

4-Butylbenzoic acid

Synonym(s):

4-butylbenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C11H14O2
CAS Number:
Molecular Weight:
178.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C11H14O2/c1-2-3-4-9-5-7-10(8-6-9)11(12)13/h5-8H,2-4H2,1H3,(H,12,13)

Inchi Key

JFKUBRAOUZEZSL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99%

Country of Origin

US

Physical Form

Solid

Melting Point

100-103C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service