Inchi Code
1S/C10H15Br/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h7-9H,1-6H2
Inchi Key
VQHPRVYDKRESCL-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Argon charged
Country of Origin
US
Physical Form
Solid
Melting Point
119C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service