Skip to Content
MilliporeSigma

ALNH9A9DB7AC

Aladdin Scientific

(Bromomethyl)cyclohexane

Synonym(s):

(bromomethyl)cyclohexane

Sign In to View Organizational & Contract Pricing.

Select a Size

5 G

$9.90

25 G

$13.90

100 G

$44.90

Vendor SKU: B121199-5G

$9.90


Ships in 5 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
177.08
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

99-100C/40mm

Inchi Code

1S/C7H13Br/c8-6-7-4-2-1-3-5-7/h7H,1-6H2

Inchi Key

UUWSLBWDFJMSFP-UHFFFAOYSA-N

Purity

99%

Country of Origin

US

Shipping Temperature

Normal

Flash Point

68C(154F)

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/B121199-SCI_079cea1cbd2515a0991901b21a6de912.pdf

signalword

Warning

pictograms

Exclamation markFlame

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service