Skip to Content
MilliporeSigma

ALNH9A9DB86B

Aladdin Scientific

6-Bromo-1-hexene

Synonym(s):

6-bromo-1-hexene

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ALNH9A9DB86B is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
163.06
MDL number:

Skip To

bp

47-51C/16mmHg

Inchi Code

1S/C6H11Br/c1-2-3-4-5-6-7/h2H,1,3-6H2

Inchi Key

RIMXEJYJXDBLIE-UHFFFAOYSA-N

Storage temp.

Store at 2-8C,Protected from light,Argon charged

Purity

95%

Country of Origin

US

Shipping Temperature

Wet ice

Flash Point

54C(129F)

Looking for similar products? Visit Product Comparison Guide

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/B122541-SCI_5292cfc816fef8c9eb7eec6598458d79.pdf

signalword

Warning

pictograms

Exclamation markFlame

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library