mp
46 C
Iupac Name
6-bromoisoquinoline
Inchi Code
1S/C9H6BrN/c10-9-2-1-8-6-11-4-3-7(8)5-9/h1-6H
Inchi Key
ZTEATMVVGQUULZ-UHFFFAOYSA-N
storage temp.
Store at 2-8C,Argon charged
Purity
97%
Country of Origin
US
Shipping Temperature
Wet ice
Product Link
https://www.aladdinsci.com/6-bromoisoquinoline-bioactive-small-molecules-by-aladdin-scientific-b134936.html
Flash Point
>110 C
Melting Point
46 C
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/B134936-SCI_aa4711c76cf6ed28e0597572bb49487e.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
