Skip to Content
MilliporeSigma

ALNH9A9DBDAD

Aladdin Scientific

1,4-Bis(3-aminopropyl)piperazine

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
200.33
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

158C

mp

15C

Iupac Name

3-[4-(3-aminopropyl)-1-piperazinyl]propylamine

Inchi Code

1S/C10H24N4/c11-3-1-5-13-7-9-14(10-8-13)6-2-4-12/h1-12H2

InChI key

XUSNPFGLKGCWGN-UHFFFAOYSA-N

storage temp.

Store at 2-8C,Argon charged

Purity

98%

Country of Origin

US

Shipping Temperature

Wet ice

Product Link

https://www.aladdinsci.com/1-4-bis-3-aminopropyl-piperazine-bioactive-small-molecules-by-aladdin-scientific-b151903.html

Flash Point

168C

Boiling point

158C

Melting Point

15C

pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/B151903-SCI_afb7e863f1a90639bc87dc5a2c1c4fc1.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service