Skip to Content
MilliporeSigma

ALNH9A9DC132

Aladdin Scientific

Butyl Propionate

Sign In to View Organizational & Contract Pricing

Select a Size

25 ML

$12.90

100 ML

$32.90

500 ML

$70.90

Vendor SKU: B152863-25ML

$12.90


Ships in 5 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
130.19
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

146 C

Iupac Name

butyl propionate

Inchi Code

1S/C7H14O2/c1-3-5-6-9-7(8)4-2/h3-6H2,1-2H3

Inchi Key

BTMVHUNTONAYDX-UHFFFAOYSA-N

Purity

99%

Country of Origin

US

Shipping Temperature

Normal

Product Link

https://www.aladdinsci.com/butyl-propionate-bioactive-small-molecules-by-aladdin-scientific-b152863.html

Flash Point

38 C

Boiling point

146 C

pictograms

Flame

Signal Word

Warning

Hazard Statements

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/B152863-SCI_02052c4cd7cc3f15bcb794d36f54aa5c.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service