Iupac Name
4-bromo-3-fluorophenylboronic acid
Inchi Code
1S/C6H5BBrFO2/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,10-11H
InChI key
ACLQPRPXJMWADE-UHFFFAOYSA-N
storage temp.
Store at 2-8C,Argon charged
Purity
97%
Country of Origin
US
Shipping Temperature
Wet ice
Product Link
https://www.aladdinsci.com/4-bromo-3-fluorophenylboronic-acid-contains-varying-amounts-of-anhydride-bioactive-small-molecules-by-aladdin-scientific-b169987.html
Signal Word
Warning
Hazard Statements
MSDS
https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/B169987-SCI_284e5a7eacc4aa0580fd3c2e4f8c66b8.pdf
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service