Skip to Content
MilliporeSigma

ALNH9A9DF100

Aladdin Scientific

(4-Bromophenyl)(morpholino)methanone

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
270.13
MDL number:

Iupac Name

4-(4-bromobenzoyl)morpholine

Inchi Code

1S/C11H12BrNO2/c12-10-3-1-9(2-4-10)11(14)13-5-7-15-8-6-13/h1-4H,5-8H2

InChI key

FTAGZJAFWFNHAP-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Product Link

https://www.aladdinsci.com/4-bromophenyl-morpholino-methanone-bioactive-small-molecules-by-aladdin-scientific-b480368.html

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/B480368-SCI_1dbf1477758adee33e639a5e1782833f.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library