Skip to Content
MilliporeSigma

ALNH9A9E63FA

Aladdin Scientific

Dicyandiamide(DCD)

Synonym(s):

N''-cyanoguanidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ALNH9A9E63FA is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C2H4N4
CAS Number:
Molecular Weight:
84.08
MDL number:

Skip To

Inchi Code

1S/C2H4N4/c3-1-6-2(4)5/h(H4,4,5,6)

Inchi Key

QGBSISYHAICWAH-UHFFFAOYSA-N

Shipping Temperature

Wet ice

Purity

98%

Storage temp.

Store at 2-8C

Country of Origin

US

Melting Point

208-211C

Physical Form

Solid

Density

1.4


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library