Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

ALNH9A9E6C37

Aladdin Scientific

cis-1,3-Dichloropropene Standard

Synonym(s):

(1E)-1,3-dichloro-1-propene

Sign In to View Organizational & Contract Pricing.

Select a Size

1 ML

$175.90

Vendor SKU: D128136-1ML

$175.90


Ships in 5 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C3H4Cl2/c4-2-1-3-5/h1-2H,3H2

Inchi Key

UOORRWUZONOOLO-UHFFFAOYSA-N

Shipping Temperature

Wet ice

Storage temp.

Store at 2-8C

Country of Origin

US

Boiling Point

104C

Density

0.7915

hstatements

signalword

Danger


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service