Skip to Content
MilliporeSigma

ALNH9A9E6C9F

Aladdin Scientific

3,4-Dihydroxybenzaldehyde

Synonym(s):

3,4-dihydroxybenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

5 G

$14.90

25 G

$23.90

100 G

$79.90

500 G

$294.90

Vendor SKU: D130075-5G

$14.90


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C7H6O3
CAS Number:
Molecular Weight:
138.12
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C7H6O3/c8-4-5-1-2-6(9)7(10)3-5/h1-4,9-10H

Inchi Key

IBGBGRVKPALMCQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Country of Origin

US

Physical Form

Solid

Melting Point

153-154C

Density

1.409

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service