Skip to Content
MilliporeSigma

ALNH9A9E7008

Aladdin Scientific

Diethyl 3,5-pyrazoledicarboxylate

Synonym(s):

diethyl 1H-pyrazole-3,5-dicarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H12O4N2
CAS Number:
Molecular Weight:
212.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H12N2O4/c1-3-14-8(12)6-5-7(11-10-6)9(13)15-4-2/h5H,3-4H2,1-2H3,(H,10,11)

Inchi Key

MBWXLICVQZUJOW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Room temperature

Country of Origin

US

Physical Form

Solid

Melting Point

55 C



Still not finding the right product?

Explore all of our products under Diethyl 3,5-pyrazoledicarboxylate


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library