Skip to Content
MilliporeSigma

ALNH9A9E9BB0

Aladdin Scientific

4-(Dimethylamino)azobenzene-4′-sulfonyl chloride

Synonym(s):

4-{(E)-[4-(dimethylamino)phenyl]diazenyl}benzenesulfonyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ALNH9A9E9BB0 is currently not available for purchase. Contact Technical Service

Skip To

Physical Form

Solid

Inchi Code

1S/C14H14ClN3O2S/c1-18(2)13-7-3-11(4-8-13)16-17-12-5-9-14(10-6-12)21(15,19)20/h3-10H,1-2H3

Inchi Key

VTVWTPGLLAELLI-UHFFFAOYSA-N

Shipping Temperature

Wet ice

Purity

95%

Storage temp.

Store at 2-8C,Protected from light,Argon charged

Country of Origin

US

Melting Point

185 C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library