Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ALNH9A9EB298

Aladdin Scientific

Eucalyptol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$28.90
500 ML
$79.90

Vendor SKU: E111233-100ML

$28.90


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
100 ML
$28.90
500 ML
$79.90

About This Item

CAS Number:
Molecular Weight:
154.25
MDL number:

Vendor SKU: E111233-100ML

$28.90


Ships in 7 days - from Aldrich Partner.

bp

176-177 C (lit.)

mp

1-2 C (lit.)

density

0.921

Iupac Name

1,3,3-trimethyl-2-oxabicyclo[2.2.2]octane

Inchi Code

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3

InChI key

WEEGYLXZBRQIMU-UHFFFAOYSA-N

Purity

99%

Country of Origin

US

Shipping Temperature

Normal

Product Link

https://www.aladdinsci.com/eucalyptol-bioactive-small-molecules-by-aladdin-scientific-e111233.html

Flash Point

48 C

Boiling point

176-177 C (lit.)

Melting Point

1-2 C (lit.)

density

0.921

pictograms

Exclamation markFlame

Signal Word

Warning

Hazard Statements

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/E111233-SCI_59295b6a6f58651321ea2f634edcec28.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service