Skip to Content
MilliporeSigma

ALNH9A9EB3A0

Aladdin Scientific

3-Ethyl-2,4-dimethylpyrrole

Sign Into View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
123.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

197C/710mmHg

Iupac Name

3-ethyl-2,4-dimethyl-1H-pyrrole

Inchi Code

1S/C8H13N/c1-4-8-6(2)5-9-7(8)3/h5,9H,4H2,1-3H3

InChI key

ZEBBLOXDLGIMEG-UHFFFAOYSA-N

storage temp.

Store at 2-8C,Argon charged

Purity

97%

Country of Origin

US

Shipping Temperature

Wet ice

Product Link

https://www.aladdinsci.com/3-ethyl-2-4-dimethylpyrrole-bioactive-small-molecules-by-aladdin-scientific-e123131.html

Flash Point

71C

Boiling point

197C/710mmHg

pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/E123131-SCI_8d93a021e97a0681d8e774c2507fa553.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service