Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

ALNH9A9EC851

Aladdin Scientific

Ethyl 3-hydrazinobenzoate hydrochloride

Synonym(s):

ethyl 3-hydrazinobenzoate hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ALNH9A9EC851 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
216.67
MDL number:

Skip To

Inchi Code

1S/C9H12N2O2.ClH/c1-2-13-9(12)7-4-3-5-8(6-7)11-10;/h3-6,11H,2,10H2,1H3;1H

Inchi Key

YMKGDNLSBACMSE-UHFFFAOYSA-N

Storage temp.

Argon charged

Purity

95%

Country of Origin

US

Shipping Temperature

Normal

Looking for similar products? Visit Product Comparison Guide

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/E479828-SCI_8e0ee315f84e0e5fc4bee396c8494de4.pdf

hstatements

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library