Skip to Content
MilliporeSigma

ALNH9A9EF9CB

Aladdin Scientific

Gallic acid monohydrate

Synonym(s):

3,4,5-trihydroxybenzoic acid hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

5 G

$11.90

25 G

$17.90

100 G

$39.90

500 G

$142.90

Vendor SKU: G104228-5G

$11.90


Ships in 5 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
188.14
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

235-240C

Inchi Code

1S/C7H6O5.H2O/c8-4-1-3(7(11)12)2-5(9)6(4)10;/h1-2,8-10H,(H,11,12);1H2

Inchi Key

IUTKPPDDLYYMBE-UHFFFAOYSA-N

Storage temp.

Argon charged

Purity

98%

Country of Origin

US

Shipping Temperature

Normal

Flash Point

250C

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/G104228-SCI_560110d0d1d1760c395e2928c9318ffa.pdf

hstatements

pcodes

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service