Skip to Content
MilliporeSigma

ALNH9A9F05D1

Aladdin Scientific

Gallic acid monohydrate

Synonym(s):

3,4,5-trihydroxybenzoic acid hydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ALNH9A9F05D1 is currently not available for purchase. Contact Technical Service

About This Item

Linear Formula:
C7H8O6
CAS Number:
Molecular Weight:
188.14
MDL number:

Skip To

Inchi Code

1S/C7H6O5.H2O/c8-4-1-3(7(11)12)2-5(9)6(4)10;/h1-2,8-10H,(H,11,12);1H2

Inchi Key

IUTKPPDDLYYMBE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Protected from light,Argon charged

Country of Origin

US

Melting Point

235-240C

Flash Point

250C

Density

1.694

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library