Skip to Content
MilliporeSigma

ALNH9A9F0BAB

Aladdin Scientific

2,2,4,4,6,8,8-Heptamethylnonane (HMN)

Synonym(s):

2,2,4,4,6,8,8-heptamethylnonane

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C16H34
CAS Number:
Molecular Weight:
226.45
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C16H34/c1-13(10-14(2,3)4)11-16(8,9)12-15(5,6)7/h13H,10-12H2,1-9H3

Inchi Key

VCLJODPNBNEBKW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

US

Physical Form

Liquid

Boiling Point

247 C

Flash Point

102C

Density

0.793


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service