Skip to Content
MilliporeSigma

ALNH9A9F2B8A

Aladdin Scientific

2-Iodo-2-methylpropane (stabilized with Copper chip)

Synonym(s):

2-iodo-2-methylpropane

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
Molecular Weight:
184.02
MDL number:

bp

99-100C

mp

-38C

Inchi Code

1S/C4H9I/c1-4(2,3)5/h1-3H3

Inchi Key

ANGGPYSFTXVERY-UHFFFAOYSA-N

Storage temp.

Store at 2-8C,Argon charged

Purity

95%

Country of Origin

US

Shipping Temperature

Wet ice

Flash Point

7C

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/I157543-SCI_774ef04c9ee2fce8bdc4eb96afd9bc89.pdf

signalword

Danger


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library