Skip to Content
MilliporeSigma

ALNH9A9F2C65

Aladdin Scientific

N,N,N′,N′-Tetramethyl-p-phenylenediamine

Synonym(s):

N~1~,N~1~,N~4~,N~4~-tetramethyl-1,4-benzenediamine

Sign In to View Organizational & Contract Pricing.

Select a Size

5 G

$31.90

10 G

$47.90

25 G

$107.90

50 G

$161.90

100 G

$307.90

Vendor SKU: I165342-5G

$31.90


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C10H16N2
CAS Number:
Molecular Weight:
164.25
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C10H16N2/c1-11(2)9-5-7-10(8-6-9)12(3)4/h5-8H,1-4H3

Inchi Key

CJAOGUFAAWZWNI-UHFFFAOYSA-N

Shipping Temperature

Ice chest + Ice pads

Purity

99%

Storage temp.

Protected from light,Store at -20C,Argon charged

Country of Origin

US

Boiling Point

260C

Melting Point

48-52C

Flash Point

110 C

Physical Form

Solid

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service