Skip to Content
MilliporeSigma

ALNH9A9F2E54

Aladdin Scientific

N-(3-Aminopropyl)piperidine

Synonym(s):

3-(1-piperidinyl)propylamine

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C8H18N2/c9-5-4-8-10-6-2-1-3-7-10/h1-9H2

Inchi Key

JMUCXULQKPWSTJ-UHFFFAOYSA-N

Storage temp.

Protected from light,Room temperature,Argon charged

Purity

95%

Country of Origin

US

Shipping Temperature

Normal

Flash Point

82.2C

hstatements

signalword

Danger

pictograms

CorrosionExclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service