Skip to Content
MilliporeSigma

ALNH9A9F2F43

Aladdin Scientific

3-iodo-4-methoxy-1H-pyrrolo[2,3-b]pyridine

Synonym(s):

3-iodo-1H-pyrrolo[2,3-b]pyridin-4-yl methyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

1 G

$433.90

Vendor SKU: I178264-1G

$433.90


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C8H7O1N2I1
CAS Number:
Molecular Weight:
274.06
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C8H7IN2O/c1-12-6-2-3-10-8-7(6)5(9)4-11-8/h2-4H,1H3,(H,10,11)

Inchi Key

PGVRYUWFBSFOBO-UHFFFAOYSA-N

Shipping Temperature

Wet ice

Purity

97%

Storage temp.

Store at 2-8C

Country of Origin

US

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service