Skip to Content
MilliporeSigma
Get 22% off for Pi Day through March 31.Save Now

ALNH9A9F33D8

Aladdin Scientific

Isododecane

Synonym(s):

2-methylundecane

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C12H26
CAS Number:
Molecular Weight:
170.34
MDL number:

Inchi Code

1S/C12H26/c1-4-5-6-7-8-9-10-11-12(2)3/h12H,4-11H2,1-3H3

Inchi Key

GTJOHISYCKPIMT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

80%

Storage temp.

Room temperature

Country of Origin

US

Boiling Point

180 C at 760 mmHg

Flash Point

42C

signalword

Danger

pictograms

Health hazardFlame

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library