Skip to Content
MilliporeSigma

ALNH9A9F36B0

Aladdin Scientific

5-Iodo-1-pentyne

Synonym(s):

5-iodo-1-pentyne

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ALNH9A9F36B0 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
194.02
MDL number:

Skip To

Inchi Code

1S/C5H7I/c1-2-3-4-5-6/h1H,3-5H2

Inchi Key

SEHFYZRHGUPLSY-UHFFFAOYSA-N

Storage temp.

Store at 2-8C,Protected from light

Purity

98%

Country of Origin

US

Shipping Temperature

Wet ice

Flash Point

8 C

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/I465322-SCI_2636a14b9303088ed0a66861bb79fe30.pdf

hstatements

signalword

Danger

pictograms

CorrosionFlame

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library