Skip to Content
MilliporeSigma

ALNH9A9F5228

Aladdin Scientific

Methyl red

Synonym(s):

2-{(E)-[4-(dimethylamino)phenyl]diazenyl}benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ALNH9A9F5228 is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
269.3
MDL number:

Skip To

mp

179 - 182 C

Inchi Code

1S/C15H15N3O2/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-10H,1-2H3,(H,19,20)

Inchi Key

CEQFOVLGLXCDCX-UHFFFAOYSA-N

Purity

Country of Origin

US

Shipping Temperature

Normal

Physical Form

Solid

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/M101206-SCI_49447d5bc4bb4cd7273da99a46a4d010.pdf

pcodes


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library