Skip to Content
MilliporeSigma

ALNH9A9F5229

Aladdin Scientific

Methyl red

Synonym(s):

2-{(E)-[4-(dimethylamino)phenyl]diazenyl}benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

25 G

$10.90

100 G

$26.90

500 G

$91.90

Vendor SKU: M101209-25G

$10.90


Ships in 5 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
269.3
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

179 - 182 C

Inchi Code

1S/C15H15N3O2/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-10H,1-2H3,(H,19,20)

Inchi Key

CEQFOVLGLXCDCX-UHFFFAOYSA-N

Purity

-

Country of Origin

US

Shipping Temperature

Normal

Looking for similar products? Visit Product Comparison Guide

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/M101209-SCI_7d29a1793160fbe4439e92e77e8f8fa8.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service