Skip to Content
MilliporeSigma

ALNH9A9F53D6

Aladdin Scientific

3-Methyl-1-pentyn-3-ol

Synonym(s):

3-methyl-1-pentyn-3-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

5 ML

$9.90

25 ML

$17.90

100 ML

$46.90

250 ML

$107.90

500 ML

$172.90

Vendor SKU: M106691-5ML

$9.90


Ships in 5 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
98.14
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

bp

122C

mp

-30.6C

Inchi Code

1S/C6H10O/c1-4-6(3,7)5-2/h1,7H,5H2,2-3H3

Inchi Key

QXLPXWSKPNOQLE-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Shipping Temperature

Normal

Flash Point

38C

Looking for similar products? Visit Product Comparison Guide

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/M106691-SCI_b8d5c295693fff1b29c2a65fa3c7a2a4.pdf

signalword

Danger


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service