Skip to Content
MilliporeSigma

ALNH9A9FAF79

Aladdin Scientific

5-Norbornen-2-ol, mixture of endo and exo

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

CAS Number:
Molecular Weight:
110.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

104-106 C

Iupac Name

bicyclo[2.2.1]hept-5-en-2-ol

Inchi Code

1S/C7H10O/c8-7-4-5-1-2-6(7)3-5/h1-2,5-8H,3-4H2

InChI key

MKOSBHNWXFSHSW-UHFFFAOYSA-N

storage temp.

Store at -20C

Purity

98%

Country of Origin

US

Shipping Temperature

Ice chest + Ice pads

Product Link

https://www.aladdinsci.com/5-norbornen-2-ol-mixture-of-endo-and-exo-bioactive-small-molecules-by-aladdin-scientific-n101933.html

Flash Point

51 C

Melting Point

104-106 C

pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/N101933-SCI_65d085193f5f748cbe763587afc5b896.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service