Skip to Content
MilliporeSigma

ALNH9AA00453

Aladdin Scientific

Phosphocholine Chloride Sodium Salt Hydrate

Synonym(s):

sodium 2-(trimethylammonio)ethyl phosphate chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H13O4N1Cl1P1Na2
CAS Number:
Molecular Weight:
263.57
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H14NO4P.ClH.2Na/c1-6(2,3)4-5-10-11(7,8)9;;;/h4-5H2,1-3H3,(H-,7,8,9);1H;;/q;;2*+1/p-2

Inchi Key

HMCRHROSIPYRGL-UHFFFAOYSA-L

Shipping Temperature

Normal

Purity

98%

Country of Origin

US

Storage temp.

Room temperature;Argon charged

Physical Form

Solid



hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library