Skip to Content
MilliporeSigma

ALNH9AA09D1C

Aladdin Scientific

1,2,4-Triazole-3-carboxamide

Synonym(s):

1H-1,2,4-triazole-5-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Product ALNH9AA09D1C is currently not available for purchase. Contact Technical Service

About This Item

CAS Number:
Molecular Weight:
112.09
MDL number:

Skip To

mp

315C

Inchi Code

1S/C3H4N4O/c4-2(8)3-5-1-6-7-3/h1H,(H2,4,8)(H,5,6,7)

Inchi Key

ZEWJFUNFEABPGL-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Shipping Temperature

Normal

MSDS

https://ald-pub-files.oss-cn-shanghai.aliyuncs.com/aladdinsci/pdp/sds/1/T162266-SCI_479bbb9c5f2386ecb3f2c4f6bc7c88d6.pdf

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library