Skip to Content
MilliporeSigma

ALNH9AA0DADE

Aladdin Scientific

2,3,4,5-Tetrahydro-1H-1,4-benzodiazepine

Synonym(s):

2,3,4,5-tetrahydro-1H-1,4-benzodiazepine

Sign In to View Organizational & Contract Pricing.

Select a Size

50 MG

$138.90

250 MG

$518.90

1 G

$1,036.90

Vendor SKU: T700685-50MG

$138.90


Ships in 5 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

mp

96-100

Inchi Code

1S/C9H12N2/c1-2-4-9-8(3-1)7-10-5-6-11-9/h1-4,10-11H,5-7H2

Inchi Key

MLXBHOCKBUILHN-UHFFFAOYSA-N

Storage temp.

Room temperature

Purity

95%

Country of Origin

US

Shipping Temperature

Normal

Physical Form

Solid

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

pictograms

CorrosionExclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service