Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day through March 27.Save Now

ALNH9AD32285

Aladdin Scientific

L-Azetidine-2-carboxylic acid

Synonym(s):

2-azetidinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

50 MG

$9.90

250 MG

$14.90

1 G

$18.90

5 G

$92.90

Vendor SKU: A124088-50MG

$9.90


Ships in 5 days - from Aldrich Partner.


About This Item

Linear Formula:
C4H7O2N1
CAS Number:
Molecular Weight:
101.11
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Physical Form

Solid

Inchi Code

1S/C4H7NO2/c6-4(7)3-1-2-5-3/h3,5H,1-2H2,(H,6,7)

Inchi Key

IADUEWIQBXOCDZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99%

Country of Origin

US

Melting Point

217C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service