Skip to Content
MilliporeSigma

ALNH9AD33EDA

Aladdin Scientific

2,5-Difluorocinnamic acid

Synonym(s):

3-(2,5-difluorophenyl)acrylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H6F2O2/c10-7-2-3-8(11)6(5-7)1-4-9(12)13/h1-5H,(H,12,13)

Inchi Key

XAWHCSKPALFWBI-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library