Skip to Content
MilliporeSigma

ALNH9AD355B3

Aladdin Scientific

4'-Hydroxychalcone

Synonym(s):

1-(4-hydroxyphenyl)-3-phenylprop-2-en-1-one

Sign In to View Organizational & Contract Pricing.

Select a Size

1 G

$9.90

5 G

$13.90

25 G

$45.90

100 G

$181.90

500 G

$682.90

Vendor SKU: H157052-1G

$9.90


Ships in 5 days - from Aldrich Partner.


Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C15H12O2/c16-14-9-7-13(8-10-14)15(17)11-6-12-4-2-1-3-5-12/h1-11,16H

Inchi Key

UAHGNXFYLAJDIN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

US

Melting Point

174 C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service