Physical Form
Solid
Density
1.195
Inchi Code
1S/C10H10O3/c1-13-9-5-3-2-4-8(9)6-7-10(11)12/h2-7H,1H3,(H,11,12)
Inchi Key
FEGVSPGUHMGGBO-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Country of Origin
US
Melting Point
183-187C
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service