Sign In to View Organizational & Contract Pricing.
Select a Size
1 ML
$88.90
Vendor SKU: M425049-1ML
$88.90
Ships in 5 days - from Aldrich Partner.
Skip To
Inchi Code
1S/C10H10O3/c1-13-9-5-3-2-4-8(9)6-7-10(11)12/h2-7H,1H3,(H,11,12)
Inchi Key
FEGVSPGUHMGGBO-UHFFFAOYSA-N
Shipping Temperature
Ice chest + Ice pads
Purity
10
Storage temp.
Store at -80C
Country of Origin
US
Melting Point
183-187C
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service