Skip to Content
MilliporeSigma

ALNH9AD36A67

Aladdin Scientific

cis-6-Nonen-1-ol

Synonym(s):

non-6-en-1-ol

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h3-4,10H,2,5-9H2,1H3

Inchi Key

XJHRZBIBSSVCEL-UHFFFAOYSA-N

Shipping Temperature

Wet ice

Purity

95%

Storage temp.

Store at 2-8C,Argon charged

Country of Origin

US

Boiling Point

112C

Flash Point

95C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service