Skip to Content
MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now

ALNH9AD37ABF

Aladdin Scientific

(1R,2R)-(-)-N-p-Tosyl-1,2-diphenylethylenediamine

Synonym(s):

N-(2-amino-1,2-diphenylethyl)-4-methylbenzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C21H22O2N2S1
CAS Number:
Molecular Weight:
366.48
MDL number:

Physical Form

Solid

Density

1.224

Inchi Code

1S/C21H22N2O2S/c1-16-12-14-19(15-13-16)26(24,25)23-21(18-10-6-3-7-11-18)20(22)17-8-4-2-5-9-17/h2-15,20-21,23H,22H2,1H3

Inchi Key

UOPFIWYXBIHPIP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

US

Melting Point

128-131C

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library