Skip to Content
MilliporeSigma

Skip To

07542

(SP)-1-Dicyclohexylphosphino-2-[(S)-α-(dimethylamino)-2-(dicyclohexylphosphino)benzyl]ferrocene

≥97%

Synonym(s):

(1R)-1-(Dicyclohexylphosphino)-2-[(S)-[2-(dicyclohexylphosphino)phenyl](dimethylamino)methyl]ferrocene (acc to CAS), Taniaphos SL-T002-2

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C43H63FeNP2
CAS Number:
Molecular Weight:
711.76
UNSPSC Code:
12352005
PubChem Substance ID:
MDL number:


assay

≥97%

optical purity

ee: ≥99%

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.CN(C)[C@H]([C]2[CH][CH][CH][C]2P(C3CCCCC3)C4CCCCC4)c5ccccc5P(C6CCCCC6)C7CCCCC7

InChI

1S/C38H58NP2.C5H5.Fe/c1-39(2)38(35-27-17-29-37(35)41(32-22-11-5-12-23-32)33-24-13-6-14-25-33)34-26-15-16-28-36(34)40(30-18-7-3-8-19-30)31-20-9-4-10-21-31;1-2-4-5-3-1;/h15-17,26-33,38H,3-14,18-25H2,1-2H3;1-5H;/t38-;;/m0../s1

InChI key

DOWXTPVXDDGHHG-ZCLNGFLVSA-N

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
H9026-5G04061833811863
118305100004022536701318
118305000104054839011498
H9026-25G04061835545674
H5269-25G04061835521852
H5269-5G04061835545551
52864-1AMP-F04061826696033
52864-6X1AMP-F04061833328521
118305010004054839012174
118305002504022536176802
07542-500MG04061832622439
07542-100MG04061832622422

Questions

Reviews

No rating value

Active Filters