Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

105929

Sigma-Aldrich

2-Ethoxyaniline

98%

Synonym(s):

o-Phenetidine, 2-Ethoxyaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4NH2
CAS Number:
Molecular Weight:
137.18
Beilstein/REAXYS Number:
637069
EC Number:
MDL number:
UNSPSC Code:
12352103

vapor pressure

1 mmHg ( 67 °C)

assay

98%

refractive index

n20/D 1.555 (lit.)

bp

231-233 °C (lit.)

density

1.051 g/mL at 25 °C (lit.)

SMILES string

CCOc1ccccc1N

InChI

1S/C8H11NO/c1-2-10-8-6-4-3-5-7(8)9/h3-6H,2,9H2,1H3

InChI key

ULHFFAFDSSHFDA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service