Documents

106429

Sigma-Aldrich

1-Tetradecanesulfonic acid sodium salt

97%

Synonym(s):
Sodium tetradecanesulfonate
Linear Formula:
CH3(CH2)13SO3Na
CAS Number:
Molecular Weight:
300.43
Beilstein/REAXYS Number:
3920141
MDL number:

assay

97%

mp

>300 °C (lit.)

SMILES string

[Na+].CCCCCCCCCCCCCCS([O-])(=O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service